missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ YPD Broth

Product Code. 10574579
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type Unit Size
10574579 500 g Bottle 500g
11738882 2 kg Pail 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10574579 Supplier BD Supplier No. 242820
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for maintaining and propagating yeasts. BD™ Difco™ YPD Broth is a dehydrated powder for preparing culture media in molecular microbiology procedures.

General methods in yeast genetics specify using yeast extract-peptone-dextrose (YPD) medium for cultivating Saccharomyces cerevisiae and other yeasts. Yeasts grow well on a minimal medium containing only dextrose and salts. The addition of protein and yeast cell extract hydrolysates allows faster growth so that during exponential or log-phase growth, the cells divide every 90 minutes.

  • Peptone acts as a source of carbon, nitrogen, vitamins and minerals
  • Yeast extract supplies B-complex vitamins, which stimulate bacterial growth
  • Dextrose is the carbohydrate source
  • pH (6.5% solution, 25°C): 6.5 ± 0.2

Order Info

Prices are for units of one each unless otherwise specified.

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Content And Storage <30°C
Form Dehydrated Powder
Media Type YPD Broth
Product Type Nutrient Broth
Formulation Yeast extract 10g/L, Peptone 20g/L, Dextrose 20g/L
For Use With (Application) For maintaining and propagating Yeasts in molecular microbiology procedures
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.