missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Tryptone Glucose Extract Agar
Click to view available options
:
Tryptone Glucose Extract Agar; Quantity: 500g
Description
Cultivation of bacteria in water, milk and other dairy products
Certifications
AOAC, COMPF
Specifications
Specifications
| Certifications/Compliance | AOAC, COMPF |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Tryptone Glucose Extract Agar |
| Product Type | Agar |
| Formulation | Beef extract 3g/L, Tryptone 5g/L, Dextrose (Glucose) 1g/L, Agar 15g/L |
| For Use With (Application) | Cultivation of a wide variety of fastidious microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction