missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Selenite Broth

Product Code. 11772864
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11772864 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11772864 Supplier BD Supplier No. 227540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For selective enrichment of Salmonella from food

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BS12, MCM9, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type Selenite Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 5g/L, Lactose 4g/L, Sodium Selenite 4g/L, Sodium Phosphate 10g/L
For Use With (Application) Selective enrichment of Salmonella from foods
Packaging Type Bottle
Quantity 500 g
compliance-icons compliance-icons
Product Identifier
  • selinite broth
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium selinite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.