missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: PALCAM Medium Base

Product Code. 11799052
Change view
Click to view available options
:
PALCAM Medium Base; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11799052 PALCAM Medium Base; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11799052 Supplier BD Supplier No. 263620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with PALCAM Antimicrobic Supplement in isolating and cultivating Listeria from foods

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF, ISO, SMD
Content And Storage <30°C
Form Dehydrated
Media Type PALCAM Medium Base
Product Type PALCAM Medium base
Formulation Columbia Blood Agar base 39g/L (Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 5g/L, Beef heart infusion from 500 g 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 15g/L); Mannitol 10g/L; Dextrose 0.5g/L; Esculin 1g/L; Ferric Ammonium Citrate 0.5g/L; Lithium Chloride 15g/L; Phenol red 0.08g/L; Acriflavine HCl 5mg/L; Polymyxin B Sulfate 0.01g/L; Agar 2g/L
For Use With (Application) For isolating and cultivating Listeria, particularly from foods and Milk products
Packaging Type Bottle
Quantity 500 g
compliance-icons compliance-icons
Product Identifier
  • Palcam Medium Base
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: lithium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.