missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Mycoplasma Broth (PPLO Broth without Crystal Violet)
Click to view available options
:
PPLO Broth without Crystal Violet; 500g
Description
- Used for isolating and cultivating Mycoplasma, when supplemented with nutritive enrichments
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Mycoplasma Broth |
| Product Type | Microbiology Media |
| Formulation | Beef Heart 6g/L, Peptone 10g/L, Sodium Chloride 5g/L |
| For Use With (Application) | Isolation and cultivation of mycoplasma, when supplemented with nutritive enrichments |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction