missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Mueller Hinton Broth

Product Code. 11703503
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
Product Code. Quantity Packaging Type unitSize
11703503 500 g Bottle 500g
11793493 2 kg Pail 2kg
2 options
This item is not returnable. View return policy

Product Code. 11703503

Brand: BD 275730

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used in the cultivation of a wide variety of fastidious and nonfastidious microorganisms. BD™ Difco™ Mueller Hinton Broth is a general purpose medium that is not cation adjusted. It may be used in antimicrobial susceptibility testing of microorganisms by broth dilution methods.

Mueller Hinton Broth, not cation-adjusted, is a dehydrated culture medium that is not supplemented with calcium or magnesium ions. It is formulated to have a low thymine and thymidine content. It may be used for broth dilution antimicrobial susceptibility testing as long as the calcium and magnesium ion concentrations are adjusted according to CLSI standard M7.

  • Acid hydrolysate (digest) of casein and beef extract supply amino acids and other nitrogenous substances, minerals, vitamins, carbon and other nutrients to support the growth of microorganisms
  • Starch acts as a protective colloid against toxic substances in the medium; hydrolysis of the starch during autoclaving provides a small amount of dextrose, which is a source of energy
  • Displays a cultural response to: Enterococcus faecalis 29212, Escherichia coli 25922, Pseudomonas aeruginosa 27853 and Staphylococcus aureus 29213
  • pH (2.1% solution, 25°C): 7.3 ± 0.1
  • Calcium: 2.9–5.9mg/L; Magnesium: 3.2–5.2mg/L
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, CLSI
Color Light beige (with a few dark specks)
Content And Storage Refrigerate
Form Dehydrated Powder
Media Type Mueller Hinton Broth
Product Type Microbiology Media
Formulation Beef extract 2g/L, Acid digest of Casein 17.5g/L, Starch 1.5g/L
For Use With (Application) Cultivation of wide variety of fastidious and non-fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.