missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: MIO Medium

Product Code. 11713483
Click to view available options
:
Motility Indole Ornithin (MIO) Medium; 500g
This item is not returnable. View return policy

Product Code. 11713483

Brand: BD 273520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to demonstrate motility, indole production and ornithine decarboxylase activity for the differentiation of Enterobacteriaceae

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM
Content And Storage Refrigerate
Form Dehydrated
Media Type MIO Medium
Product Type Motility Test Medium
Formulation Yeast extract 3g/L, Peptone 10g/L, Tryptone 10g/L, L-Ornithine HCl 5g/L, Dextrose 1g/L, Agar 2g/L, Bromcresol Purple 0.02g/L
For Use With (Application) Demonstrate motility, indole production and Ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.