Learn More
BD Difco™ Dehydrated Culture Media: Middlebrook 7H10 Agar
Use with Glycerol and Middlebrook OADC Enrichment for isolation and cultivation of mycobacteria. BD Difco™ Dehydrated Culture Media: Middlebrook 7H10 Agar is part of a large selection of dehydrated media.
Brand: BD 262710
Additional Details : Weight : 0.01000kg
Description
- Dehydrated
- Culture Media
- Middlebrook 7H10 Agar
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
ISO, 9000, EP
Specifications
ISO, 9000, EP | |
Dehydrated | |
Agar | |
Isolation of Escherichia coli 25922, Mycobacterium tuberculosis H37Ra 25177, Mycobacterium kansasii, group I 12478, Mycobacterium scrofulaceum, group II 19981, Mycobacterium intracellulare, group III 13950, Mycobacterium fortuitum, group IV 6841 | |
500 g |
<30°C | |
Middlebrook 7H10 Agar | |
Ammonium Sulfate 0.5g/900mL, Monopotassium Phosphate 1.5g/L,/900mL, Disodium Phosphate 1.5 g/900mL, Sodium Citrate 0.4g/900mL, Magnesium Sulfate 25mg/900mL, Calcium Chloride 0.5mg/900mL, Zinc Sulfate 1mg/900mL, Copper Sulfate 1mg/900mL, L-Glutamic Acid (sodium salt) 0.5g/900mL, Ferric Ammonium Citrate 0.04g/900mL, Pyridoxine Hydrochloride 1mg/900mL, Biotin 0.5mg/900mL, Malachite green 250μg/900mL, Agar 15g/900mL | |
Bottle |