missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: McClung Toabe Agar Base
Click to view available options
:
McClung Toabe Agar Base; 500g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | McClung Toabe Agar Base |
| Product Type | Agar |
| Formulation | Proteose Peptone 40g/L, Dextrose 2g/L, Disodium Phosphate 5g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 2g/L, Magnesium Sulfate 0.1g/L, Agar 25g/L |
| For Use With (Application) | Egg Yolk enrichment 50% for isolating and detecting Clostridium perfringens in food based on the Lecithinase reaction |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction