missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Marine Agar 2216

Product Code. 11718183
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11718183 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11718183 Supplier BD Supplier No. 212185

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolation, cultivation and enumeration of heterotrophic marine bacteria. BD Difco™ Dehydrated Culture Media: Marine Agar 2216 is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Marine Agar 2216

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Marine Agar 2216
Product Type Agar
Formulation Peptone 5g/L, Yeast extract 1g/L, Ferric Citrate 0.1g/L, Sodium Chloride 19.45g/L, Magnesium Chloride 8.8g/L, Sodium Sulfate 3.24g/L, Calcium Chloride 1.8g/L, Potassium Chloride 0.55g/L, Sodium Bicarbonate 0.16g/L, Potassium Bromide 0.08g/L, Strontium Chloride 34mg/L, Boric Acid 22mg/L, Sodium Silicate 4mg/L, Sodium Fluoride 2.4mg/L, Ammonium Nitrate 1.6mg/L, Disodium Phosphate 8mg/L, Agar 15g/L
For Use With (Application) Isolation, cultivation and enumeration of heterotrophic marine bacteria
Packaging Type Bottle
Quantity 500 g
compliance-icons
Product Identifier
  • marine broth 2216
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: calcium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.