missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Malt Extract Broth
Click to view available options
:
Malt Extract Broth; 500g
Description
The use of malt and malt extracts for the propagation of yeasts and molds is quite common. Malt Extract Broth is recommended for the examination of yeasts and molds in the U.S. Food and Drug Administration’s Bacteriological Analytical Manual.
- Contains malt extract which provides the carbon, protein, and nutrient sources required for growth of microorganisms.
- Maltose is added as an energy source, and Dextrose is included as a source of fermentable carbohydrate.
- Yeast extract provides the vitamins and cofactors required for growth and additional sources of nitrogen and carbon.
- Provides good recovery of Aspergillus brasiliensis (niger), Candida albicans and Saccharomyces cerevisiae.
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
CE, BAM
Specifications
Specifications
| Certifications/Compliance | CE, BAM |
| Color | Light beige |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Malt Extract Broth |
| Product Type | Microbiology Media |
| Formulation | Malt extract 6g/L, Maltose, Technical 1.8g/L, Dextrose 6g/L, Yeast extract 1.2g/L |
| For Use With (Application) | For Isolation, detection and enumeration of yeasts and molds |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction