missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: m TGE Broth
Click to view available options
:
m Tryptone Glucose. Extract (TGE) Broth; 500g
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, COMPF |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | m Tryptone Glucose Extract (TGE) Broth |
| Product Type | Microbiology Media |
| Formulation | Beef extract 6g/L, Tryptone 10g/L, Dextrose (Glucose) 2g/L |
| For Use With (Application) | Isolation of Thermophilic bacteria |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction