missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: m LES Endo Agar

Product Code. 11743483
Click to view available options
Quantity:
100 g
500 g
For Use With (Application):
Coliform enumeration
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11743483

Brand: BD 273620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For enumerating coliforms in water by membrane filtration

  • Formulation: Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g

Certifications

CE, COMPF, SMD, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, COMPF, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type mLES Endo Agar
Product Type Agar
Formulation Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g/L, Basic Fuchsin 0.8g/L, Agar 15g/L
For Use With (Application) Coliform enumeration
Packaging Type Bottle
Quantity 500 g
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : basic fuchsin, sodium sulphite
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.