missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Lactobacilli Broth AOAC
Click to view available options
:
Lactobacilli Broth AOAC; 100g
Specifications
Specifications
| Certifications/Compliance | AOAC |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Lactobacilli Broth |
| Product Type | Microbiology Media |
| Formulation | Peptonized Milk 15g/L, Yeast extract 5g/L, Dextrose 10g/L, Tomato Juice 5g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 1g/L |
| For Use With (Application) | AOAC used to prepare microorganisms inocula for Vitamin assays |
| Packaging Type | Bottle |
| Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction