missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: ISP Medium 4

Product Code. 11753503
Click to view available options
:
International Streptomyces Project (ISP) Medium 4; 500g
This item is not returnable. View return policy

Product Code. 11753503

Brand: BD 277210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

International Streptomyces Project Inorganic Salts Starch Agar

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage Refrigerate
Form Dehydrated
Media Type ISP-Yeast Malt Agar (Medium 4)
Product Type Agar
Formulation Soluble Starch 10g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate USP 1g/L, Sodium Chloride 1g/L, Ammonium Sulfate 2g/L, Calcium Carbonate 2g/L, Ferrous Sulfate 1mg/L, Manganous Chloride 1mg/L, Zinc Sulfate 1mg/L, Agar 20g/L
For Use With (Application) For characterizing Streptomyces species according to the International Streptomyces project
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.