missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: EY Tellurite Enrichment
Click to view available options
:
EY Tellurite Enrichment (use with Baird-Parker Agar Base); BD Difco; 100mL
Description
Used with Baird-Parker Agar Base for the isolation of S. aureus
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Additive | EY Tellurite Enrichment |
| Form | Viscous |
| For Use With (Application) | Baird Parker Agar base |
| Packaging Type | Bottle |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction