missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Columbia Broth
Click to view available options
:
Columbia Broth; 500g
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Columbia Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Proteose Peptone No. 3 5g/L, Tryptic digest of Beef heart 3g/L, L-Cysteine HCl 0.1g Dextrose 2.5g Sodium Chloride 5g/L, Magnesium Sulfate (anhydrous) 0.1g Ferrous Sulfate 0.02g/L, Sodium Carbonate 0.6g/L, Tris (Hydroxymethyl) Aminomethane 0.83g/L, Tris (Hydroxymethyl) Aminomethane HCl 2.86g/L |
| For Use With (Application) | Cultivation of fastidious microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction