missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Columbia Blood Agar Base
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, EP, ISO |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Columbia Blood Agar Base |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 5g/L, Beef heart infusion from 500 g 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?