missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Columbia Blood Agar Base
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, EP, ISO |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Columbia Blood Agar Base |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 5g/L, Beef heart infusion from 500 g 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms |
| Packaging Type | Pail |
| Quantity | 2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction