missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Buffered Peptone Water Preenrichment

Product Code. p-4902036
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11748273

Brand: BD 218105

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For injured Salmonella in food

Certifications

ISO/DIS, 11014, BAM, CCAM, ISO, USDA

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BAM, CCAM, ISO, USDA
Content And Storage <30°C
Form Dehydrated
Media Type Peptone Water
Product Type Peptone Water
Formulation Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
For Use With (Application) Isolation of Salmonella species
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.