missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Brain Heart Infusion Agar
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Dimensione della confezione:
2kg
500g
Descrizione
BHI Agar has proven to be effective in the cultivation of a wide variety of fastidious microorganisms, including many types of pathogens, fungi and yeasts. Also used with added antibiotics in isolating fungi. It derives its nutrients from the brain heart infusion, peptone and dextrose components.
- The peptones and infusion are sources of organic nitrogen, carbon, sulfur, vitamins and trace substances
- Dextrose is a carbohydrate source that microorganisms utilize by fermentative action
- The medium is buffered through the use of disodium phosphate
- To prepare solution, suspend 52g in 1L purified water
- pH (5.2% solution, 25°C): 7.4 ± 0.2
- Good cultural response with: Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615
Specifica
Specifica
| Certifications/Compliance | ISO 9000, AOAC, BAM, CCAM, COMPF, EPA, MCM9, SMD, SMWW, USDA |
| Color | Beige |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | BHI Agar |
| Product Type | Agar |
| Formulation | Calf brain infusion from 200g 7.7g/L, Beef heart infusion from 250g 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 15g/L |
| For Use With (Application) | Isolation of Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615 |
| Packaging Type | Bottle |
| Quantity | 500 g |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto