missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: BG Sulfa Agar Brilliant Green
Click to view available options
:
Brilliant Green (BG) Sulfa Agar; 500g
Specifications
Specifications
| Certifications/Compliance | CCAM, COMPF, USDA |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Brilliant Green Sulfa Agar |
| Product Type | Agar |
| Formulation | Yeast extract 3g/L, Proteose Peptone 10g/L, Lactose 10g/L, Sodium Sulfapyridine 1g/L, Sodium Chloride 5g/L, Agar 20g/L, Brilliant green 12.5mg/L, Phenol red 0.08g/L |
| For Use With (Application) | For enriching Salmonella prior to isolation procedure |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction