missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Antibiotic Medium 2
Description
- Penassay Seed Agar Microbiological assay of antibiotics
Specifications
Specifications
| Certifications/Compliance | AOAC, USP |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Antibiotic Medium 2 |
| Product Type | Antibiotic Medium |
| Formulation | Beef extract 1.5g/L, Yeast extract 3g/L, Peptone 6g/L, Agar 15g/L |
| For Use With (Application) | Penassay base Agar Microbiological assay of Antibiotics |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?