missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Actinomycete Isolation Agar
Click to view available options
:
Actinomycete Isolation Agar; 500g
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Actinomycete Isolation Agar |
| Product Type | Agar |
| Formulation | Sodium Caseinate 2g/L, Asparagine 0.1g/L, Sodium Propionate 4g/L, Dipotassium Phosphate 0.5g/L, Magnesium Sulfate 0.1g/L, Ferrous Sulfate 1mg/L, Agar 15g/L |
| For Use With (Application) | For isolating and cultivating Actinomycetes from soil and Water |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Signal Word
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : sodium propionate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction