Learn More
BD Salmonella and Shigella Typing Antisera
Product Code. 17392361
Change view
Click to view available options
Type:
Salmonella O Antiserum Group A Factors 1, 2, 12
Salmonella O Antiserum Group B Factors 1, 4, 12, 27
Salmonella O Antiserum Group B Factors 1, 4, 5, 12
Salmonella O Antiserum Group C1 Factors 6, 7
Salmonella O Antiserum Group C2 Factors 6, 8
Salmonella O Antiserum Group C3 Factors 8
Salmonella O Antiserum Group E2 Factors 3, 15
Salmonella O Antiserum Group E3 Factors (3), (15), 34
Salmonella O Antiserum Group G Factors 13, 22, 23, (36), (37)
Salmonella O Antiserum Group I Factor 16
Salmonella O Antiserum Group K Factor 18
Salmonella O Antiserum Poly A-I Factors 1-16
Shigella Antiserum Poly Group C
Shigella Poly Group C1
Shigella Poly Group C2
Unit Size:
3mL
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17392361 Salmonella O Antiserum Group C3 Factors 8 Each
17332361 Salmonella O Antiserum Group I Factor 16 Each
11774985 Salmonella O Antiserum Poly A-I Factors 1-16 3mL
11712804 Salmonella O Antiserum Group K Factor 18 3mL
11712874 Shigella Poly Group C1 3mL
11722874 Shigella Poly Group C2 3mL
11762894 Shigella Antiserum Poly Group C 3mL
11735035 Salmonella O Antiserum Group B Factors 1, 4, 5, 12 3mL
11912805 Salmonella O Antiserum Group A Factors 1, 2, 12 3mL
17362361 Salmonella O Antiserum Group C1 Factors 6, 7 Each
11795965 Salmonella O Antiserum Group C2 Factors 6, 8 3mL
17382361 Salmonella O Antiserum Group B Factors 1, 4, 12, 27 Each
17372361 Salmonella O Antiserum Group E2 Factors 3, 15 Each
17302371 Salmonella O Antiserum Group E3 Factors (3), (15), 34 Each
17312371 Salmonella O Antiserum Group G Factors 13, 22, 23, (36), (37) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 17392361 Supplier BD Supplier No. 230161
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Clia Complexity High
Detectable Analytes N. gonorrhoeae
Type Salmonella O Antiserum Group C3 Factors 8
Quantity 3 mL
Form Lyophilized
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.