missing translation for 'onlineSavingsMsg'
Learn More

BD Salmonella and Shigella Typing Antisera

Product Code. 11774985
Change view
Click to view available options
Type:
Salmonella O Antiserum Group A Factors 1, 2, 12
Salmonella O Antiserum Group B Factors 1, 4, 12, 27
Salmonella O Antiserum Group B Factors 1, 4, 5, 12
Salmonella O Antiserum Group C1 Factors 6, 7
Salmonella O Antiserum Group C2 Factors 6, 8
Salmonella O Antiserum Group C3 Factors 8
Salmonella O Antiserum Group E2 Factors 3, 15
Salmonella O Antiserum Group E3 Factors (3), (15), 34
Salmonella O Antiserum Group G Factors 13, 22, 23, (36), (37)
Salmonella O Antiserum Group I Factor 16
Salmonella O Antiserum Group K Factor 18
Salmonella O Antiserum Poly A-I Factors 1-16
Shigella Antiserum Poly Group C
Shigella Poly Group C1
Shigella Poly Group C2
Unit Size:
3mL
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11774985 Salmonella O Antiserum Poly A-I Factors 1-16 3mL
17332361 Salmonella O Antiserum Group I Factor 16 Each
11712804 Salmonella O Antiserum Group K Factor 18 3mL
11712874 Shigella Poly Group C1 3mL
11722874 Shigella Poly Group C2 3mL
11762894 Shigella Antiserum Poly Group C 3mL
11735035 Salmonella O Antiserum Group B Factors 1, 4, 5, 12 3mL
11912805 Salmonella O Antiserum Group A Factors 1, 2, 12 3mL
17362361 Salmonella O Antiserum Group C1 Factors 6, 7 Each
11795965 Salmonella O Antiserum Group C2 Factors 6, 8 3mL
17382361 Salmonella O Antiserum Group B Factors 1, 4, 12, 27 Each
17372361 Salmonella O Antiserum Group E2 Factors 3, 15 Each
17392361 Salmonella O Antiserum Group C3 Factors 8 Each
17302371 Salmonella O Antiserum Group E3 Factors (3), (15), 34 Each
17312371 Salmonella O Antiserum Group G Factors 13, 22, 23, (36), (37) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 11774985 Supplier BD Supplier No. 222641
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Clia Complexity High
Detectable Analytes Salmonella
Type Salmonella O Antiserum Poly A-I Factors 1-16
Quantity 3 mL
Form Lyophilized
compliance-icons
Product Identifier
  • vial sal O antiserum poly grp A-I & Vi Lyo
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.