missing translation for 'onlineSavingsMsg'
Learn More

BD D- Luciferin Sodium Salt

Product Code. 15838788
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15838788

Brand: BD 556875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Sodium salt

Specific for firefly luciferase in the illustrated reactions

Purified by HPLC techniques to remove dehydroluciferin and L-Luciferin, both of which are competitive inhibitors of firefly luciferase. Form dehydroluciferin in presence of ultraviolet light and/or moisture

  • Synthetically made substrate
  • lyophilized, in monosodium salt form
  • Non-buffered solution
  • Reconstitute lyophilized D-Luciferin with deionized water

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized
Kit Contents Sodium salt
Type D-Luciferin
Quantity 50 mg
Content And Storage -20°C
For Use With (Application) Luciferase assay

Research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.