missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
- Protein concentration of 100,000pg/ml (when reconstituted in 0.4mL Assay Diluent)
- Regulatory Status: RUO
- Recombinant
Flow cytometry, controls.
Order Info
100 tests.
Specifications
Specifications
| Buffer | Aqueous Buffered Solution containing BSA and ProClin™ 150 |
| Includes | Technical data sheet |
| Kit Size | 100 Tests |
| Product Type | Cytometric Ab Control Standards |
| For Use With (Application) | Flow cytometry |
| Regulatory Status | RUO |
| Protein Concentration | 100000pg/mL |
| Storage Requirements | 4°C |
| Volume (Metric) | 0.2mL |
| Species | Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?