missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mouse IL-10 Enhanced Sensitivity Flex Set

Product Code. 15821769
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15821769

Brand: BD 562263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bead-based immunoassay capable of measuring mouse Interleukin-10 in serum and cell culture supernatant sample

Contains one vial each of Enhanced Sensitivity Capture Bead and Detection Reagent (Part A) and two vials of Standard. The Enhanced Sensitivity Detection Reagent (Part B) is provided in the CBA Mouse Enhanced Sensitivity Master Buffer Kit. The Enhanced Sensitivity Capture Bead and Detection Reagent (Part A) components of this flex set have been formulated to a 20× concentration to ensure product performance when multiplexed.

  • Capable of measuring soluble analytes present in very low concentrations
  • Must be used in conjunction with a CBA Mouse Enhanced Sensitivity Master Buffer Kit, a flow cytometer, and the FCAP Array Software

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-10, flow cytometry
Product Type Mouse IL-10 enhanced sensitivity flex set (bead C4)
compliance-icons
Product Identifier
  • Mouse IL-10 Enhanced Sensitivity Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.