missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Contains lyophilized recombinant human monocyte chemoattractant protein-1
- When reconstituted in 0.4mL assay diluent, has a protein concentration of 25,000pg/mL
Specifications
Specifications
| Buffer | Aqueous Buffered Solution containing BSA and ProClin™ 150 |
| Includes | Technical data sheet |
| Kit Size | 100 Tests |
| Product Type | Cytometric Ab Control Standards |
| For Use With (Application) | Flow cytometry |
| Regulatory Status | RUO |
| Protein Concentration | 25000pg/mL |
| Storage Requirements | 4°C |
| Volume (Metric) | 0.2mL |
| Form | Lyophilized |
Safety and Handling
Product Identifier
- Human MCP-1 Standard
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: CMIT/MIT
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?