missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Flex Set Standard

Product Code. 15889398
Click to view available options
Description:
Human IFN-gamma Flex Set Standard
Human IL-10 Flex Set Standard
Human IL-1beta Flex Set Standard
Human IL-2 Flex Set Standard
Human IL-4 Flex Set Standard
Human IL-6 Flex Set Standard
Human IL-8 Flex Set Standard
Human TNF Flex Set Standard
Mouse IL-10 Flex Set Standard
Mouse IL-6 Flex Set Standard
Mouse TNF Flex Set Standard
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15889398

Brand: BD 558456

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as standard control

This product contains lyophilized recombinant human Interferon-λ (IFN-λ)

  • Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Description Human IFN-gamma Flex Set Standard
Format Lyophilized
Includes Lyophilized recombinant Human Interferon-l (IFN-l)
Quantity 0.2 mL
Regulatory Status RUO
Product Type Cytometric Bead Array (CBA) Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.