missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Rat TNF; Flex Set, Bead C8
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Rat TNF; Standard; Rat TNF; PE Detection Reagent; Rat TNF; Capture Bead C8
The BD CBA Rat TNF Flex Set is a bead-based immunoassay capable of measuring rat tumor necrosis factor (TNF) in serum and cell culture supernatant samples. Rat and mouse reactivity was determined by testing samples with the BD CBA Rat TNF Flex Set.
- Reactivity: Rat
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 10000pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | TNF |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Sample |
| Content And Storage | 4°C |
| For Use With (Application) | Flow cytometry, for detecting Rat tumor necrosis factor |
| Product Type | Rat TNF flex set (bead C8) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction