missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IL-23 p19/p40 Flex Set (Bead D7)

Product Code. 15587135
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Product Code. 15587135

Brand: BD 562575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Includes

  • Kit contents:
  • Capture Bead D7: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • PE Detection Reagent: stored in aqueous buffered solution containing BSA and ≤0.09% sodium azide
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150

Measures mouse Interleukin-23 p19/p40 (IL-23 p19/p40) in serum and cell culture supernatant samples

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: mouse
  • Top standard point: 2500pg/mL

Flow Cytometry/Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-23
Cross Reactivity Mouse
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Measures Mouse interleukin-23 (p19/p40), flow cytometry
Product Type Mouse IL-23 p19/p40 flex set (bead D7)
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Mouse IL-23 p19/p40 Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.