missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse IL-17A; Flex Set, Bead C5
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Mouse IL-17A; Standard; Mouse IL-17A; PE Detection Reagent; Mouse IL-17A; Capture Bead C5
The BD CBA Mouse IL-17A Flex Set is a bead-based immunoassay capable of measuring mouse IL-17A in serum and cell culture supernatant samples
- Reactivity: QC Tested: Mouse
- Bead Position: C5
- Limit of detection is 0.95pg/mL
With BD CBA Mouse/Rat soluble protein master buffer kit (Cat. No. 558266, 100 tests, or 558267, 500 tests), a flow cytometer, and the FCAP Array™ software
Specifications
Specifications
| Antigen | IL-17A |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-17A, standard, Mouse IL-17A, PE detection reagent, Mouse IL-17A, capture bead C5 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse IL-17A flex set (bead C5) |
Safety and Handling
Product Identifier
- BD Cytometric Bead Array (CBA) Mouse IL-17A Flex Set
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : CMIT/MIT
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction