missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse IL-1β Enhanced Sensitivity Flex Set (Bead B9)
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
- Kit contents:
- Enhanced Sensitivity Capture Bead B9: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
- Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
- Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Mouse Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
- Bead-based immunoassay
- Reactivity: mouse
- Standard assay range: 274 to 200000fg/mL
Flow Cytometry/Immunoassay
Specifications
Specifications
| Antigen | IL-1beta |
| Cross Reactivity | Mouse |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Measures Mouse interleukin-1beta, flow cytometry |
| Product Type | Mouse IL-1beta enhanced sensitivity flex set (bead B9) |
Safety and Handling
Product Identifier
- Flex Set
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : CMIT/MIT
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction