missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse IFN-γ Enhanced Sensitivity Flex Set ,Bead A4

Product Code. 15881739
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15881739

Brand: BD 562233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Mouse IFN-γ Standard, Mouse IFN-γ Enhanced Sensitivity Capture Bead A4, Mouse IFN-γ Detection Reagent (Part A)

Sufficient for 100 tests

Host Species:
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-gamma
Cross Reactivity Mouse
Includes Mouse IFN-g standard, Mouse IFN-g enhanced sensitivity capture bead A4, Mouse IFN-g detection reagent (part A)
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse IFN-gamma enhanced sensitivity flex set (bead A4)
compliance-icons
Product Identifier
  • BD Cytometric Bead Array (CBA) Mouse IFN-
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.