missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse G-CSF; Flex Set, Bead D4
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Mouse G-CSF; Standard; Mouse G-CSF; PE Detection Reagent; Mouse G-CSF; Capture Bead D4
- Capable of measuring mouse Granulocyte-Colony Stimulating Factor (G-CSF) in serum and cell culture supernatant samples
Specifications
Specifications
| Antigen | G-CSF |
| Cross Reactivity | Mouse |
| Includes | Mouse G-CSF, standard, Mouse G-CSF, PE detection reagent, Mouse G-CSF, capture bead D4 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse G-CSF flex set (bead D4) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction