missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human MIP-1α; Flex Set, Bead B9

Product Code. 15819308
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819308 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819308 Supplier BD Supplier No. 558325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human MIP-1α; Standard; Human MIP-1α; PE Detection Reagent; Human MIP-1α; Capture Bead B9

For detecting human Macrophage Inflammatory Protein-1α in serum, plasma, and cell culture samples

The BD CBA Human MIP-1α Flex Set is a bead-based immunoassay capable of measuring human Macrophage Inflammatory Protein-1α (MIP-1α), also known as CCL3, in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human MIP-1α Flex Set.

  • Reactivity: Human
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1alpha
Cross Reactivity Human
Includes Human MIP-1a, standard, Human MIP-1a, PE detection reagent, Human MIP-1a, capture bead B9
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human MIP-1alpha flex set (bead B9)
compliance-icons
Product Identifier
  • Human MIP-1 alpha Flex Set
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT mixture

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.