missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human MCP-1; Flex Set, Bead D8

Product Code. 15899278
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15899278

Brand: BD 558287

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Human MCP-1; Standard; Human MCP-1; PE Detection Reagent; Human MCP-1; Capture Bead D8

For detecting human monocyte chemoattractant proteins

The BD CBA Human MCP-1 Flex Set is a bead-based immunoassay capable of measuring human monocyte chemoattractant protein-1 (MCP-1), also known as CCL2, in serum, plasma, and cell culture supernatant samples. Human and non-human primate reactivity was determined by testing samples with the BD CBA Human MCP-1 Flex Set

  • Bead position: D8
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Cross Reactivity Human
Includes Human MCP-1, standard, Human MCP-1, PE detection reagent, Human MCP-1, capture bead D8
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human MCP-1 flex set (bead D8)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.