missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit Standard controls
Description
- Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
- Should be reconstituted and serially diluted before use
Human Inflammatory Cytokine Standards
- Contain mixture of recombinant human Interleukin (IL)-8, IL-1β, IL-6, IL-10, Tumor Necrosis Factor (TNF), and IL-12p70
- Use with BD™ Cytometric Bead Array (CBA) Human Inflammatory Cytokine Kit
- Size: 0.2mL
- Contain mixture of recombinant human Interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF)
- Use with BD™ Cytometric Bead Array (CBA) Human Th1/Th2/TH17 Cytokine Kit
- Size: 1 vial
Flow Cytometry
Specifications
Specifications
| Cross Reactivity | Human |
| Includes | Mixture of recombinant Human interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, interferon-g (IFN-g) and Tumor Necrosis Factor (TNF) |
| Quantity | 1 Vial |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human Th1/Th2/Th17 Cytokine Standards |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?