missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IL-8; Flex Set, Bead A9
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human IL-8; Standard; Human IL-8; PE Detection Reagent; Human IL-8; Capture Bead A9
Reactivity
- QC Testing: Human
- Testing in Development: Non-human Primate
Flow cytometry
Specifications
Specifications
| Antigen | IL-8 |
| Cross Reactivity | Human |
| Includes | Human IL-8, standard, Human IL-8, PE detection reagent, Human IL-8, capture bead A9 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-8 flex set (bead A9) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction