missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IL-21; Flex Set, Bead B8
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human IL-21; Standard; Human IL-21; PE Detection Reagent; Human IL-21; Capture Bead B8
The BD CBA Human IL-21 Flex Set is a bead-based immunoassay capable of measuring human Interleukin 21 (IL-21) in serum, plasma, and cell culture supernatant samples
- Reactivity: QC Tested: Human
- Bead Position: B8
- Limit of detection: 34.3pg/mL
With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), a flow cytometer, and the FCAP Array™ software
Specifications
Specifications
| Antigen | IL-21 |
| Cross Reactivity | Human |
| Includes | Human IL-21, standard, Human IL-21, PE detection reagent, Human IL-21, capture bead B8 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-21 flex set (bead B8) |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction