missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human IL-17A Standard

Product Code. 15831219
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15831219 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15831219 Supplier BD Supplier No. 561668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lyophilized recombinant Human Interleukin-17A (IL-17A) protein

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Approximates the CBA activity of Human IL-17A at a concentration of 2500 pg/mL when reconstituted in 4.0 mL assay diluent
  • Quantity: 100 tests

Flow cytometry/ Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes 1 Vial, lyophilized, Aqueous Buffered, reconstitute to 4mL
Quantity 100 Tests
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry, Immunoassay
Product Type Human IL-17A standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.