missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IL-11; Flex Set, Bead D5
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human IL-11; Standard; Human IL-11; PE Detection Reagent; Human IL-11; Capture Bead D5
Human reactivity was determined by testing samples with the BD CBA Human IL-11 Flex Set.
Specifications
Specifications
| Antigen | IL-11 |
| Cross Reactivity | Human |
| Includes | Capture bead, detection reagent and standard |
| Quantity | 100 Tests |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-11 flex set (bead D5) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction