missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IgA; Flex Set, Bead C9
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human Ig, Standard (two vials), Human total IgA PE Detection Reagent (one vial), Human total IgA Capture Bead C9 (one vial)
- Human reactivity was determined by testing samples with IgA Flex Set
- Theoretical limit of detection is 7.35 ng/mL
- Storage buffer: Aqueous buffered solution containing BSA along with other reagents depending on kit components
Flow cytometry
Specifications
Specifications
| Antigen | IgA |
| Cross Reactivity | Human |
| Includes | Human Ig, standard (2 vials), Human total IgA PE detection reagent (one vial), Human total IgA capture bead C9 (one vial) |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IgA flex set (bead C9) |
Research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction