missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human IFN-γ Enhanced Sensitivity Flex Set (Bead B8)
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Kit contents:
- Enhanced Sensitivity Capture Bead B8: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
- Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
- Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
- Bead-based immunoassay
- Reactivity: human
- Standard assay range: 274 to 200000fg/mL
Flow cytometry/Immunoassay
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Human |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Measures Human interferon-gamma, flow cytometry and immunoassay |
| Product Type | Human IFN-gamma enhanced sensitivity flex set (bead B8) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction