missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human GM-CSF; Flex Set, Bead C9
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human GM-CSF; Standard; Human GM-CSF; PE Detection Reagent; Human GM-CSF; Capture Bead C9
The BD CBA Human GM-CSF Flex Set is a bead-based immunoassay capable of measuring human Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human GM-CSF Flex Set.
- Reactivity: Human
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | GM-CSF |
| Cross Reactivity | Human |
| Includes | Human GM-CSF, standard, Human GM-CSF, PE detection reagent, Human GM-CSF, capture bead C9 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human GM-CSF flex set (bead C9) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction