missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit

Product Code. 15871099
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15871099

Brand: BD 561523

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Enhanced Sensitivity Detection Reagent (Part B), Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector

Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A), provided with the Flex Set and Enhanced Sensitivity Detection Reagent (Part B), provided in Master Buffer Kit with all other supporting reagents. Buffers, detection reagent and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.

  • Analyte working assay range: 274 to 200000fg/mL
  • Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
  • Available in pack sizes of 100 and 500 tests

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes Assay diluent, capture bead Diluent, detection reagent diluent, Wash Buffer, enhanced sensitivity detection reagent (Part B), instrument setup beads (A1, A9, F1, F9), PE instrument setup bead F1, PE positive control detector
Quantity 500 Tests
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry
Product Type Human enhanced sensitivity master Buffer kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.