missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit

Product Code. 15851099
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
Each
Denne vare kan ikke returneres. Se returpolitik

Produktkode 15851099

missing translation for 'mfr': BD 561521

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Indeholder

Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Enhanced Sensitivity Detection Reagent (Part B), Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector

Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

Detection reagents used in BD™ CBA Human Enhanced Sensitivity Flex Set System consist of two parts, Analyte Specific Detection Reagent (Part A), provided with the Flex Set and Enhanced Sensitivity Detection Reagent (Part B), provided in Master Buffer Kit with all other supporting reagents. Buffers, detection reagent and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.

  • Analyte working assay range: 274 to 200000fg/mL
  • Should not be used with Flex Sets from any other BD CBA System such as soluble protein, cell signalling etc
  • Available in pack sizes of 100 and 500 tests

Flow Cytometry

TRUSTED_SUSTAINABILITY

Tekniske data

Cross Reactivity Human
Includes Assay diluent, capture bead Diluent, detection reagent diluent, Wash Buffer, enhanced sensitivity detection reagent (Part B), instrument setup beads (A1, A9, F1, F9), PE instrument setup bead F1, PE positive control detector
Quantity 100 Tests
Content And Storage 2°C to 8°C
For Use With (Application) Flow Cytometry
Product Type Human enhanced sensitivity master Buffer kit
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.