missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Human Basic FGF; Flex Set, Bead C5
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Description
Includes
Human Basic FGF; Standard; Human Basic FGF; PE Detection Reagent; Human Basic FGF; Capture Bead C5
The BD CBA Human Basic FGF Flex Set is bead-based immunoassay capable of measuring human basic Fibroblast Growth Factor (FGF), also known as FGF-2, in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human basic FGF Flex Set.
- Reactivity: Human
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | FGF |
| Cross Reactivity | Human |
| Includes | Human basic FGF, standard, Human basic FGF, PE detection reagent, Human basic FGF, capture bead C5 |
| Quantity | 100 Tests |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human basic FGF flex set (bead C5) |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction